N'-[1-Amino-2-(2-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058713-500MG
058713-1G
101915-528EA
284.62
USD
101915-528
101915-530
N'-[1-Amino-2-(2-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(2-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₈ClN₃O₂ MW: 283.76 g/mol |
MDL Number:
MFCD10568195 CAS Number: 1053656-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...