N'-[1-Amino-2-(2-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058710-500MG
058710-1G
101915-516EA
284.62
USD
101915-516
101915-518
N'-[1-Amino-2-(2-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(2-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₈ClN₃O₃ MW: 299.76 g/mol |
MDL Number:
MFCD10568189 CAS Number: 1053655-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...