5-Amino-2-chloropyridine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-amino-2-chloropyridine-4-carboxylic acid, 5-Amino-2-chloroisonicotinic acid, 3-Amino-6-chloroisonicotinic acid

058709-1G 058709-5G
101915-512EA 44.62 USD
101915-512 101915-514
5-Amino-2-chloropyridine-4-carboxylic acid ≥95%
5-Amino-2-chloropyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD06659499
CAS Number: 58483-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR