N'-[1-Amino-1-(3-chloro-4-fluorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058708-500MG
058708-1G
101915-508EA
284.62
USD
101915-508
101915-510
N'-[1-Amino-1-(3-chloro-4-fluorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(3-chloro-4-fluorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₂H₁₅ClFN₃O₂ MW: 287.72 g/mol |
MDL Number:
MFCD10568190 CAS Number: 1053655-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...