N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058707-500MG
058707-1G
101915-504EA
284.62
USD
101915-504
101915-506
N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₄H₁₅F₆N₃O₂ MW: 371.28 g/mol |
MDL Number:
MFCD10568191 CAS Number: 1053655-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...