N'-[1-Amino-2-(3,5-bis-(trifluoromethyl)phenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058706-500MG 058706-1G
101915-500EA 284.62 USD
101915-500 101915-502
N'-[1-Amino-2-(3,5-bis-(trifluoromethyl)phenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(3,5-bis-(trifluoromethyl)phenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₅H₁₇F₆N₃O₂
MDL Number: MFCD10568198
CAS Number: 1053656-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR