2-Aminobenzo[d]thiazole-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminobenzothiazole-6-carboxylic acid

058704-1G 058704-5G 058704-10G
101915-488EA 353.85 USD
101915-488 101915-490 101915-492
2-Aminobenzo[d]thiazole-6-carboxylic acid ≥95%
2-Aminobenzo[d]thiazole-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂S
MDL Number: MFCD00054180
CAS Number: 93-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR