2-Aminobenzothiazole-6-carbonitrile ≥95%

Supplier: Matrix Scientific

058703-1G 058703-5G 058703-10G
101915-482EA 378.46 USD
101915-482 101915-484 101915-486
2-Aminobenzothiazole-6-carbonitrile ≥95%
2-Aminobenzothiazole-6-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃S
MW: 175.2104 g/mol
MDL Number: MFCD02664359
CAS Number: 19759-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR