4-Acetoxytetrahydrothiophene-3-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

058697-1G 058697-5G
101915-460EA 267.69 USD
101915-460 101915-462
4-Acetoxytetrahydrothiophene-3-carboxylic acid methyl ester ≥95%
4-Acetoxytetrahydrothiophene-3-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₈H₁₂O₄S
MDL Number: MFCD15142796
CAS Number: 1208081-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR