tert-Butyl-2-(3-{[(benzyloxy)carbonyl]amino}-5-methyl-1H-pyrazol-1-yl)benzoate ≥95%

Supplier: Matrix Scientific

058692-500MG 058692-1G
101915-434EA 1192.31 USD
101915-434 101915-436
tert-Butyl-2-(3-{[(benzyloxy)carbonyl]amino}-5-methyl-1H-pyrazol-1-yl)benzoate ≥95%
tert-Butyl-2-(3-{[(benzyloxy)carbonyl]amino}-5-methyl-1H-pyrazol-1-yl)benzoate

Hazard: Irritant

Formula: C₂₃H₂₅N₃O₄
MW: 407.47 g/mol
MDL Number: MFCD18157648
CAS Number: 1261079-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR