2-[4-(2-Methylpropoxy)phenyl]ethan-1-ol ≥95%

Supplier: Matrix Scientific

058685-500MG 058685-1G 058685-5G
101915-402EA 401.54 USD
101915-402 101915-404 101915-406
2-[4-(2-Methylpropoxy)phenyl]ethan-1-ol ≥95%
2-[4-(2-Methylpropoxy)phenyl]ethan-1-ol

Hazard: Irritant

Formula: C₁₂H₁₈O₂
MDL Number: MFCD14582731
CAS Number: 500547-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR