Methyl-1-(2-formylphenyl)-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
058670-500MG
058670-1G
101915-332EA
992.31
USD
101915-332
101915-334
Methyl-1-(2-formylphenyl)-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-1-(2-formylphenyl)-5-methyl-1H-pyrazole-3-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₃ |
MDL Number:
MFCD18157647 CAS Number: 1261079-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...