1-(4-Fluorophenyl)-2-[2-(methylsulfanyl)pyrimidin-4-yl]ethan-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Fluorophenyl)-2-(2-(methylthio)pyrimidin-4-yl)ethanone

058662-500MG 058662-1G 058662-5G
101915-296EA 598.46 USD
101915-296 101915-298 101915-300
1-(4-Fluorophenyl)-2-[2-(methylsulfanyl)pyrimidin-4-yl]ethan-1-one ≥95%
1-(4-Fluorophenyl)-2-[2-(methylsulfanyl)pyrimidin-4-yl]ethan-1-one

Hazard: Irritant

Formula: C₁₃H₁₁FN₂OS
MW: 262.31 g/mol
MDL Number: MFCD18157689
CAS Number: 217661-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR