4-(4-Aminophenoxy)-N-methylpicolinamide ≥95%
Supplier: Matrix Scientific
058611-1G
058611-5G
101915-106EA
315.38
USD
101915-106
101915-108
4-(4-Aminophenoxy)-N-methylpicolinamide ≥95%
4-(4-Aminophenoxy)-N-methylpicolinamide
Hazard: Irritant
Formula:
C₁₃H₁₃N₃O₂ |
MDL Number:
MFCD08692047 CAS Number: 284462-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...