3-Piperazin-1-yl-1,2-benzisoxazole ≥95%
Supplier: Matrix Scientific
058606-250MG
058606-1G
101915-092EA
95.38
USD
101915-092
101915-094
3-Piperazin-1-yl-1,2-benzisoxazole ≥95%
3-Piperazin-1-yl-1,2-benzisoxazole
Hazard: Irritant
Formula:
C₁₁H₁₃N₃O MW: 203.25 g/mol |
MDL Number:
MFCD08060968 CAS Number: 87691-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...