Diethyl cyclohexane-1,1-dicarboxylate ≥95%

Supplier: Matrix Scientific

058556-1G 058556-5G
101915-046EA 38.46 USD
101915-046 101915-048
Diethyl cyclohexane-1,1-dicarboxylate ≥95%
Diethyl cyclohexane-1,1-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
MDL Number: MFCD02093457
CAS Number: 1139-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR