Methyl-4-ethynylbenzoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-ethynylbenzoate
058541-1G
058541-5G
101915-038EA
67.69
USD
101915-038
101915-040
Methyl-4-ethynylbenzoate ≥97%
Methyl 4-ethynylbenzoate
Hazard: Irritant
Formula:
C₁₀H₈O₂ |
MDL Number:
MFCD00168820 CAS Number: 3034-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...