2-Aminopyrazine ≥98%

Supplier: Matrix Scientific

Synonyms: Aminopyrazine, Pyrazin-2-amine

058540-25G 058540-100G
101915-034EA 23.08 USD
101915-034 101915-036
2-Aminopyrazine ≥98%
2-Aminopyrazine

Hazard: Irritant

Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 118-121°C
MDL Number: MFCD00006137
CAS Number: 5049-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR