Methyl-2-(3,4,5-trimethoxyphenyl)acetate ≥95%
Supplier: Matrix Scientific
058507-500MG
058507-1G
101914-974EA
264.62
USD
101914-974
101914-976
Methyl-2-(3,4,5-trimethoxyphenyl)acetate ≥95%
Methyl-2-(3,4,5-trimethoxyphenyl)acetate
Hazard: Irritant
Formula:
C₁₂H₁₆O₅ Boiling Pt: 172…182 °C (10 Torr) Melting Pt: 40…41 °C |
MDL Number:
MFCD00017207 CAS Number: 2989-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...