Ethyl-1-(3-nitropyridin-2-yl)piperidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058501-500MG 058501-1G
101914-946EA 173.85 USD
101914-946 101914-948
Ethyl-1-(3-nitropyridin-2-yl)piperidine-3-carboxylate ≥95%
Ethyl-1-(3-nitropyridin-2-yl)piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₄
MW: 279.3 g/mol
MDL Number: MFCD13875962
CAS Number: 952934-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR