6-Methylpicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-2-pyridinecarboxylic acid, 6-Methylpyridine-2-carboxylic acid

058455-5G 058455-25G
101914-840EA 41.54 USD
101914-840 101914-842
6-Methylpicolinic acid ≥95%
6-Methylpicolinic acid

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00023481
CAS Number: 934-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR