(2-Amino-6-methoxyphenyl)(phenyl)methanone

Supplier: Matrix Scientific

053867-500MG 053867-2.5G
101905-284EA 423.08 USD
101905-284 101905-286
(2-Amino-6-methoxyphenyl)(phenyl)methanone
(2-Amino-6-methoxyphenyl)(phenyl)methanone

Hazard: Irritant

Formula: C₁₄H₁₃NO₂
MDL Number: MFCD13562950
CAS Number: 461694-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR