(2-Amino-6-methoxyphenyl)(2-fluorophenyl)methanone
Supplier: Matrix Scientific
053866-500MG
053866-2.5G
101905-280EA
364.62
USD
101905-280
101905-282
(2-Amino-6-methoxyphenyl)(2-fluorophenyl)methanone
(2-Amino-6-methoxyphenyl)(2-fluorophenyl)methanone
Hazard: Irritant
Formula:
C₁₄H₁₂FNO₂ |
MDL Number:
MFCD13562949 CAS Number: 1219979-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...