(2-Aminophenyl)(2-fluorophenyl)methanone
Supplier: Matrix Scientific
053865-500MG
053865-2.5G
101905-276EA
461.54
USD
101905-276
101905-278
(2-Aminophenyl)(2-fluorophenyl)methanone
(2-Aminophenyl)(2-fluorophenyl)methanone
Hazard: Irritant
Formula:
C₁₃H₁₀FNO |
MDL Number:
MFCD00007714 CAS Number: 1581-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...