6,7-Dihydrobenzo[b]thiophen-4-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-Dihydro-4-benzo[b]thiophenone, 4-Keto-4,5,6,7-tetrahydrothianaphthene

053860-1G 053860-5G
101905-272EA 38.46 USD
101905-272 101905-274
6,7-Dihydrobenzo[b]thiophen-4-one ≥95%
6,7-Dihydrobenzo[b]thiophen-4-one

Hazard: Irritant

Formula: C₈H₈OS
MW: 152.211 g/mol
MDL Number: MFCD00005861
CAS Number: 13414-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR