2-Chloro-6-ethoxybenzoic acid
Supplier: Matrix Scientific
053857-500MG
053857-2.5G
101905-260EA
500
USD
101905-260
101905-262
2-Chloro-6-ethoxybenzoic acid
2-Chloro-6-ethoxybenzoic acid
Hazard: Irritant
Formula:
C₉H₉ClO₃ |
MDL Number:
MFCD11156593 CAS Number: 1020998-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...