2-Bromo-6-ethoxybenzoic acid

Supplier: Matrix Scientific

053855-500MG 053855-2.5G
101905-256EA 364.62 USD
101905-256 101905-258
2-Bromo-6-ethoxybenzoic acid
2-Bromo-6-ethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₉BrO₃
MDL Number: MFCD13562946
CAS Number: 1220035-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR