(4-Methoxyphenyl)(4-methylphenyl)methanamine

Supplier: Matrix Scientific

053853-500MG 053853-2.5G
101905-248EA 364.62 USD
101905-248 101905-250
(4-Methoxyphenyl)(4-methylphenyl)methanamine
(4-Methoxyphenyl)(4-methylphenyl)methanamine

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD09812085
CAS Number: 161110-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR