(4-Methylphenyl)(phenyl)methanamine
Supplier: Matrix Scientific
Synonyms:
Phenyl(p-tolyl)methanamine
053852-500MG
053852-2.5G
101905-244EA
276.92
USD
101905-244
101905-246
(4-Methylphenyl)(phenyl)methanamine
(4-Methylphenyl)(phenyl)methanamine
Hazard: Irritant
Formula:
C₁₄H₁₅N MW: 197.28 g/mol |
MDL Number:
MFCD00134171 CAS Number: 55095-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...