2-(2-Benzoyl-6-chloro-1H-indol-3-yl)acetic acid
Supplier: Matrix Scientific
053850-500MG
053850-2.5G
101905-236EA
364.62
USD
101905-236
101905-238
2-(2-Benzoyl-6-chloro-1H-indol-3-yl)acetic acid
2-(2-Benzoyl-6-chloro-1H-indol-3-yl)acetic acid
Hazard: Irritant
Formula:
C₁₇H₁₂ClNO₃ |
MDL Number:
MFCD13562945 CAS Number: 231292-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...