2-Benzyl-1H-benzimidazole-6-carboxylic acid
Supplier: Matrix Scientific
053848-500MG
053848-2.5G
101905-228EA
364.62
USD
101905-228
101905-230
2-Benzyl-1H-benzimidazole-6-carboxylic acid
2-Benzyl-1H-benzimidazole-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₂N₂O₂ |
MDL Number:
MFCD13562944 CAS Number: 162400-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...