2-Benzyl-1-methyl-1H-benzimidazole-6-carboxylic acid
Supplier: Matrix Scientific
053847-500MG
053847-2.5G
101905-224EA
364.62
USD
101905-224
101905-226
2-Benzyl-1-methyl-1H-benzimidazole-6-carboxylic acid
2-Benzyl-1-methyl-1H-benzimidazole-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₄N₂O₂ |
MDL Number:
MFCD13562943 CAS Number: 193013-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...