2-Ethyl-1-methyl-1H-benzimidazole-6-carboxylic acid

Supplier: Matrix Scientific

053845-500MG 053845-2.5G
101905-220EA 364.62 USD
101905-220 101905-222
2-Ethyl-1-methyl-1H-benzimidazole-6-carboxylic acid
2-Ethyl-1-methyl-1H-benzimidazole-6-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD13562942
CAS Number: 1219960-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR