2-(2-Pyridinyl)-1H-benzimidazole-6-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2-(Pyridin-2-yl)-1H-benzo[d]imidazole-6-carboxylic acid

053844-500MG 053844-2.5G
101905-216EA 364.62 USD
101905-216 101905-218
2-(2-Pyridinyl)-1H-benzimidazole-6-carboxylic acid
2-(2-Pyridinyl)-1H-benzimidazole-6-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉N₃O₂
MW: 239.24 g/mol
MDL Number: MFCD06245483
CAS Number: 669070-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR