1,3,3-Trimethyl-6-azabicyclo[3.2.1]octane

Supplier: Matrix Scientific

053841-500MG 053841-2.5G
101905-208EA 360 USD
101905-208 101905-210
1,3,3-Trimethyl-6-azabicyclo[3.2.1]octane
1,3,3-Trimethyl-6-azabicyclo[3.2.1]octane

Hazard: Irritant

Formula: C₁₀H₁₉N
MDL Number: MFCD00066934
CAS Number: 53460-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR