1-Methyl-3-phenyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

053838-500MG 053838-2.5G
101905-196EA 364.62 USD
101905-196 101905-198
1-Methyl-3-phenyl-1H-indole-2-carboxylic acid
1-Methyl-3-phenyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₂
MDL Number: MFCD13562939
CAS Number: 91606-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR