Ethyl-4-amino-1H-imidazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 4-amino-1H-imidazole-5-carboxylate

053833-500MG 053833-2.5G
101905-180EA 384.62 USD
101905-180 101905-182
Ethyl-4-amino-1H-imidazole-5-carboxylate
Ethyl-4-amino-1H-imidazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD06858453
CAS Number: 21190-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR