2-Allyl-6-nitroaniline
Supplier: Matrix Scientific
053828-500MG
053828-2.5G
101905-164EA
364.62
USD
101905-164
101905-166
2-Allyl-6-nitroaniline
2-Allyl-6-nitroaniline
Hazard: Irritant
Formula:
C₉H₁₀N₂O₂ |
MDL Number:
MFCD13562932 CAS Number: 154490-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...