3-(4-Methylphenyl)cyclohexanone

Supplier: Matrix Scientific

053821-500MG 053821-2.5G
101905-136EA 364.62 USD
101905-136 101905-138
3-(4-Methylphenyl)cyclohexanone
3-(4-Methylphenyl)cyclohexanone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD13562928
CAS Number: 99495-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR