3-(2-Fluorophenyl)cyclohexanone
Supplier: Matrix Scientific
053819-500MG
053819-2.5G
101905-128EA
364.62
USD
101905-128
101905-130
3-(2-Fluorophenyl)cyclohexanone
3-(2-Fluorophenyl)cyclohexanone
Hazard: Irritant
Formula:
C₁₂H₁₃FO |
MDL Number:
MFCD13562926 CAS Number: 141632-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...