3-Phenylcyclohexanone ≥95%
Supplier: Matrix Scientific
053818-500MG
053818-2.5G
101905-124EA
384.62
USD
101905-124
101905-126
3-Phenylcyclohexanone ≥95%
3-Phenylcyclohexanone
Hazard: Irritant
Formula:
C₁₂H₁₄O MW: 174.239 g/mol |
MDL Number:
MFCD00209998 CAS Number: 20795-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...