3-(tert-Butyl)cyclohexanone

Supplier: Matrix Scientific

053816-500MG 053816-2.5G
101905-116EA 346.15 USD
101905-116 101905-118
3-(tert-Butyl)cyclohexanone
3-(tert-Butyl)cyclohexanone

Hazard: Irritant

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00962367
CAS Number: 936-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR