3-Benzylcyclohexanone
Supplier: Matrix Scientific
053815-500MG
053815-2.5G
101905-112EA
364.62
USD
101905-112
101905-114
3-Benzylcyclohexanone
3-Benzylcyclohexanone
Hazard: Irritant
Formula:
C₁₃H₁₆O Boiling Pt: 142…144 °C (5 Torr) |
MDL Number:
MFCD13562925 CAS Number: 85450-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...