6-(Benzyloxy)nicotinic acid

Supplier: Matrix Scientific

053812-500MG 053812-2.5G
101905-100EA 149.23 USD
101905-100 101905-102
6-(Benzyloxy)nicotinic acid
6-(Benzyloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
MDL Number: MFCD09730715
CAS Number: 94084-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR