5-Ethoxy-2-pyridine carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-Ethoxypicolinic acid

053809-500MG 053809-2.5G
101905-088EA 307.69 USD
101905-088 101905-090
5-Ethoxy-2-pyridine carboxylic acid
5-Ethoxy-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD13562922
CAS Number: 98353-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR