3-(4-Methoxyphenyl)pyrrolidine hydrochloride

Supplier: Matrix Scientific

053805-500MG 053805-2.5G
101905-076EA 384.62 USD
101905-076 101905-078
3-(4-Methoxyphenyl)pyrrolidine hydrochloride
3-(4-Methoxyphenyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD03701253
CAS Number: 91246-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR