3-(1-Naphthyl)pyrrolidine
Supplier: Matrix Scientific
053803-500MG
053803-2.5G
101905-068EA
364.62
USD
101905-068
101905-070
3-(1-Naphthyl)pyrrolidine
3-(1-Naphthyl)pyrrolidine
Hazard: Irritant
Formula:
C₁₄H₁₅N |
MDL Number:
MFCD13562919 CAS Number: 178483-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...