6-Methoxy-1,2,3,4-tetrahydroisoquinoline
Supplier: Matrix Scientific
053802-500MG
053802-2.5G
101905-064EA
169.23
USD
101905-064
101905-066
6-Methoxy-1,2,3,4-tetrahydroisoquinoline
6-Methoxy-1,2,3,4-tetrahydroisoquinoline
Hazard: Irritant
Formula:
C₁₀H₁₃NO MW: 163.2163 g/mol |
MDL Number:
MFCD01717129 CAS Number: 42923-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...