4-(3-Methyl-5-isoxazolyl)piperidine hydrochloride

Supplier: Matrix Scientific

053800-500MG 053800-2.5G
101905-056EA 769.23 USD
101905-056 101905-058
4-(3-Methyl-5-isoxazolyl)piperidine hydrochloride
4-(3-Methyl-5-isoxazolyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₅ClN₂O
MDL Number: MFCD22415177
CAS Number: 1401425-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR