3-(3-Methyl-5-isoxazolyl)piperidine hydrochloride

Supplier: Matrix Scientific

053799-500MG 053799-2.5G
101905-052EA 384.62 USD
101905-052 101905-054
3-(3-Methyl-5-isoxazolyl)piperidine hydrochloride
3-(3-Methyl-5-isoxazolyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₅ClN₂O
MDL Number: MFCD22041638
CAS Number: 1374659-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR